Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isonicotinaldehyde ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

134615-100G 134615-25G 134615-500G
BT134615-100GEA 103 USD InStock
BT134615-100G BT134615-25G BT134615-500G
Isonicotinaldehyde ≥97%
Isonicotinaldehyde
CAS: 872-85-5; EC No: 212-832-3; MDL No: MFCD00006425; RTECS: NR9400000 Liquid; Molecular Formula: C6H5NO; MW: 107.11 Melting Point: -4°; Boiling Point: 198°; Flash point: 75°C (167°F) Density (g/mL): 1.137; Refractive Index: 1.544

  • RTECS: NR9400000
  • Refractive index: 1.544
  • Liquid


Formula: C₆H₅NO
MW: 107.11 g/mol
Boiling Pt: 198 °C
Melting Pt: –4 °C
Density: 1.137 g/mL
Flash Pt: 75 °C (167 °F)
MDL Number: MFCD00006425
CAS Number: 872-85-5
EINECS: 212-832-3
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty