Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydroxypivalic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

134775-250G 134775-50G
BT134775-250GEA 219 USD InStock
BT134775-250G BT134775-50G
Hydroxypivalic acid ≥98%
Hydroxypivalic acid
CAS: 4835-90-9; EC No: 225-419-8; MDL No: MFCD00059953 Crystalline; Molecular Formula: C5H10O3; MW: 118.13

  • Crystalline


Formula: C₅H₁₀O₃
MDL Number: MFCD00059953
CAS Number: 4835-90-9
EINECS: 225-419-8
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty