Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indazole ≥99%

Supplier: BEANTOWN CHEMICAL - BTC

134790-1G 134790-25G 134790-5G
BT134790-1GEA 32 USD InStock
BT134790-1G BT134790-25G BT134790-5G
Indazole ≥99%
Indazole
CAS: 271-44-3; EC No: 205-978-4; MDL No: MFCD00005691; RTECS: NK7745000 Powder; Molecular Formula: C7H6N2; MW: 118.14 Melting Point: 145-148°

  • RTECS: NK7745000
  • Powder


Formula: C₇H₆N₂
Melting Pt: 145…148 °C
MDL Number: MFCD00005691
CAS Number: 271-44-3
EINECS: 205-978-4
Order Now
Specification Test Results
Assay 99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty