Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Polyethylene glycol 10,000

Supplier: BEANTOWN CHEMICAL - BTC

134860-1KG 134860-250G
BT134860-1KGEA 94.6 USD InStock
BT134860-1KG BT134860-250G
Polyethylene glycol 10,000
Polyethylene glycol 10,000
CAS: 25322-68-3; EC No: 203-473-3; MDL No: MFCD00081839; RTECS: TQ4110000 Flakes; Linear Formula: H(OCH2CH2)nOH; MW ≈10,000 Melting Point: 63-65°; Flash point: 265°C (509°F)

  • RTECS: TQ4110000
  • Flakes


Formula: H(OCH₂CH₂)nOH
MW: 10000 g/mol
Melting Pt: 63
Flash Pt: 265 °C (509 °F)
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 203-473-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty