Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-5-Amino-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

135720-25G 135720-5G
BT135720-25GEA 167 USD InStock
BT135720-25G BT135720-5G
(S)-5-Amino-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid ≥98%
(S)-5-Amino-2-((tert-butoxycarbonyl)amino)-5-oxopentanoic acid
CAS: 13726-85-7; EC No: 237-296-8; MDL No: MFCD00065571 Powder; Molecular Formula: C10H18N2O5 ; MW: 246.26 Melting Point: 113-116° (decomposes) Optical Rotation: [α]20/D -3.5°, c = 1 in ethanol

  • Optical rotation: [α]20/D -3.5°, c = 1 in ethanol
  • Powder


Formula: C₁₀H₁₈N₂O₅
MW: 246.3 g/mol
Melting Pt: 113…116 °C (dec.)
MDL Number: MFCD00065571
CAS Number: 13726-85-7
EINECS: 237-296-8
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty