Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tin(II) tartrate hydrate ≥95%

Supplier: BEANTOWN CHEMICAL - BTC

136475-100G 136475-25G
BT136475-100GEA 384 USD InStock
BT136475-100G BT136475-25G
Tin(II) tartrate hydrate ≥95%
Tin(II) tartrate hydrate
CAS: 815-85-0; EC No: 212-427-1; MDL No: MFCD00054358; RTECS: EJ9892500 Powder; Molecular Formula: C4H4O6Sn·xH2O; MW: 266.78 (anhydrous) Air Sensitive

  • RTECS: EJ9892500
  • Air Sensitive
  • Powder


Formula: C₄H₄O₆Sn·xH₂O
MW: 266.78 g/mol (anhy)
MDL Number: MFCD00054358
CAS Number: 815-85-0
EINECS: 212-427-1
Order Now
Specification Test Results
Assay 95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty