Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Nitrophthalic acid ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

136925-25G 136925-50G 136925-5G
BT136925-25GEA 106 USD InStock
BT136925-25G BT136925-50G BT136925-5G
4-Nitrophthalic acid ≥97%
4-Nitrophthalic acid
CAS: 610-27-5; EC No: 210-215-3; MDL No: MFCD00007252 Powder; Molecular Formula: C8H5NO6 ; MW: 211.13 Melting Point: 159-161°

  • Powder


Formula: C₈H₅NO₆
Melting Pt: 159…161 °C
MDL Number: MFCD00007252
CAS Number: 610-27-5
EINECS: 210-215-3
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty