Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sebacic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

137015-1KG 137015-250G
BT137015-1KGEA 80.88 USD InStock
BT137015-1KG BT137015-250G
Sebacic acid ≥98%
Sebacic acid
CAS: 111-20-6; EC No: 203-845-5; MDL No: MFCD00004440; RTECS: VS0875000 Powder; Linear Formula: HOOC(CH2)8COOH; MW: 202.25 Melting Point: 133-137°; Boiling Point: 295°/100 mmHg; Flash point: 220°C (428°F) Density (g/mL): 1.210

  • RTECS: VS0875000
  • Powder


Formula: C₁₀H₁₈O₄
MW: 202.25 g/mol
Boiling Pt: 374
Melting Pt: 133…137 °C
Density: 1.210 g/mL
Flash Pt: 220 °C (428 °F)
MDL Number: MFCD00004440
CAS Number: 111-20-6
EINECS: 203-845-5
Merck Index: 12,08558
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty