Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium bromide ≥99.0% ACS

Supplier: BEANTOWN CHEMICAL - BTC

137120-100G 137120-25G 137120-500G
BT137120-100GEA 29.12 USD InStock
BT137120-100G BT137120-25G BT137120-500G
Ammonium bromide ≥99.0% ACS
Ammonium bromide
CAS: 12124-97-9; EC No: 235-183-8; MDL No: MFCD00011419; RTECS: BO9155000 Granular; Linear Formula: NH4Br; MW: 97.97 Melting Point: 452° Density (g/mL): 2.43 Hygroscopic

  • RTECS: BO9155000
  • Hygroscopic
  • Granular


Formula: H₄BrN
MW: 97.94 g/mol
Melting Pt: 542
Density: 2.43 g/mL
MDL Number: MFCD00011419
CAS Number: 12124-97-9
EINECS: 235-183-8
Merck Index: 13,00505
Order Now
Specification Test Results
Assay 99.0% min.

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty