Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Noradamantanecarboxylic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

137275-1G 137275-250MG 137275-5G
BT137275-1GEA 91 USD InStock
BT137275-1G BT137275-250MG BT137275-5G
3-Noradamantanecarboxylic acid ≥98%
3-Noradamantanecarboxylic acid
CAS: 16200-53-6; MDL No: MFCD00213463 Solid; Molecular Formula: C10H14O2; MW: 166.22 Melting Point: 105-107°

  • Solid


Formula: C₁₀H₁₄O₂
Melting Pt: 105…107 °C
MDL Number: MFCD00213463
CAS Number: 16200-53-6
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty