Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromobiphenyl (PBB No. 1) ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

137310-1G 137310-25G 137310-5G
BT137310-1GEA 18 USD InStock
BT137310-1G BT137310-25G BT137310-5G
2-Bromobiphenyl (PBB No. 1) ≥98%
2-Bromobiphenyl (PBB No. 1)
CAS: 2052-07-5; EC No: 218-141-3; MDL No: MFCD00000065 Liquid; Molecular Formula: C12H9Br ; MW: 233.10 Melting Point: 1-2°; Boiling Point: 297-298°; Flash point: 110°C (230°F) Density (g/mL): 1.352; Refractive Index: 1.628

  • Refractive index: 1.628
  • Liquid


Formula: C₁₂H₉Br
MW: 233.11 g/mol
Boiling Pt: 297…298 °C
Melting Pt: 1…2 °C
Density: 1.352 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00000065
CAS Number: 2052-07-5
EINECS: 218-141-3
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty