Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride

Supplier: BEANTOWN CHEMICAL - BTC

137870-1G 137870-250MG
BT137870-1GEA 416 USD InStock
BT137870-1G BT137870-250MG
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride
[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) dichloride
CAS: 95408-45-0; MDL No: MFCD08064219 Crystalline; Molecular Formula: C26H44Cl2FeP2Pd; MW: 651.77 Melting Point: 203-208°

  • Crystalline


Formula: C₂₆H₄₄Cl₂FeP₂Pd
Melting Pt: 203…208 °C
MDL Number: MFCD08064219
CAS Number: 95408-45-0
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty