Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenol red ACS

Supplier: BEANTOWN CHEMICAL - BTC

137970-100G 137970-25G 137970-5G
BT137970-100GEA 124.2 USD InStock
BT137970-100G BT137970-25G BT137970-5G
Phenol red ACS
Phenol red
CAS: 143-74-8; EC No: 205-609-7; MDL No: MFCD00003552; RTECS: SJ7490000 Crystalline; Molecular Formula: C19H14O5S; MW: 354.38 Melting Point: 300°

  • RTECS: SJ7490000
  • Crystalline


Formula: C₁₉H₁₄O₅S
MW: 354.38 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00003552
CAS Number: 143-74-8
EINECS: 205-609-7
Merck Index: 13,07329
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty