Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Palladium ≥99.99% (trace metals basis), wire Ø 0.5 mm

Supplier: BEANTOWN CHEMICAL - BTC

138200-250CM 138200-50CM
BT138200-250CMEA 1209.32 USD InStock
BT138200-250CM BT138200-50CM
Palladium ≥99.99% (trace metals basis), wire Ø 0.5 mm
Palladium
CAS: 7440-05-3; EC No: 231-115-6; MDL No: MFCD00011167; RTECS: RT3480500 Solid; Molecular Formula: Pd; MW: 106.42 Melting Point: 1554°; Boiling Point: 2970° Density (g/mL): 12.02

  • RTECS: RT3480500
  • Solid


Formula: Pd
Boiling Pt: 2970 °C
Melting Pt: 1554 °C
Density: 12.02 g/mL
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Order Now
Specification Test Results
Assay 99.99%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty