Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-methoxynicotinate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

138255-10G 138255-250G 138255-50G
BT138255-10GEA 69 USD InStock
BT138255-10G BT138255-250G BT138255-50G
Methyl-6-methoxynicotinate ≥98%
Methyl-6-methoxynicotinate
CAS: 26218-80-4; MDL No: MFCD00674130 Solid; Molecular Formula: C8H9NO3; MW: 167.16 Melting Point: 48-53°

  • Solid


Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 48…53 °C
MDL Number: MFCD00674130
CAS Number: 26218-80-4
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty