Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Salicylonitrile ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

138540-100G 138540-25G 138540-5G
BT138540-100GEA 402 USD InStock
BT138540-100G BT138540-25G BT138540-5G
Salicylonitrile ≥98%
Salicylonitrile
CAS: 611-20-1; EC No: 210-259-3; MDL No: MFCD00002145 Solid; Molecular Formula: C7H5NO; MW: 119.12 Melting Point: 92-95°; Boiling Point: 149°/14 mmHg

  • Solid


Formula: C₇H₅NO
MW: 119.12 g/mol
Boiling Pt: 273
Melting Pt: 92…95 °C
MDL Number: MFCD00002145
CAS Number: 611-20-1
EINECS: 210-259-3
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty