Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dextrin

Supplier: BEANTOWN CHEMICAL - BTC

139135-100G 139135-500G
BT139135-100GEA 42.57 USD InStock
BT139135-100G BT139135-500G
Dextrin
Dextrin
CAS: 9004-53-9; EC No: 232-675-4; MDL No: MFCD00081554; RTECS: HH9450000 Powder Hygroscopic

  • RTECS: HH9450000
  • Hygroscopic
  • Powder


Formula: (C₆H₁₂O₆)n
MW: 180.16 g/mol
MDL Number: MFCD00081554
CAS Number: 9004-53-9
EINECS: 232-675-4
Merck Index: 13,02971
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty