Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-iodopyridine ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

139330-25G 139330-5G
BT139330-25GEA 141 USD InStock
BT139330-25G BT139330-5G
5-Bromo-2-iodopyridine ≥98%
5-Bromo-2-iodopyridine
CAS: 223463-13-6; MDL No: MFCD00691506 Crystalline; Molecular Formula: C5H3BrIN ; MW: 283.89 Melting Point: 113-117° Light Sensitive

  • Light Sensitive
  • Crystalline


Formula: C₅H₃BrIN
MW: 283.89 g/mol
Melting Pt: 113…117 °C
MDL Number: MFCD00691506
CAS Number: 223463-13-6
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty