Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Barium carbonate ≥99.95%

Supplier: BEANTOWN CHEMICAL - BTC

139670-10G 139670-250G 139670-50G
BT139670-10GEA 35.17 USD InStock
BT139670-10G BT139670-250G BT139670-50G
Barium carbonate ≥99.95%
Barium carbonate
CAS: 513-77-9; EC No: 208-167-3; MDL No: MFCD00003448; RTECS: CQ8000000 Powder/Lump; Linear Formula: BaCO3; MW: 197.35

  • RTECS: CQ8000000
  • Powder/Lump


Formula: BaCO₃
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1,III
Order Now
Specification Test Results
Assay 99.95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty