Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Polyethylene oxide, M.W. ∼1000000

Supplier: BEANTOWN CHEMICAL - BTC

139870-100G 139870-25G 139870-500G
BT139870-100GEA 78 USD InStock
BT139870-100G BT139870-25G BT139870-500G
Polyethylene oxide, M.W. ∼1000000
Polyethylene oxide
CAS: 25322-68-3; EC No: 500-038-2; MDL No: MFCD00147971 Powder; Linear Formula: (-CH2CH2O-)n; MW ≈1,000,000 Boiling Point: 200°

  • Powder


Formula: (-CH₂CH₂O-)n
Boiling Pt: >200 °C
MDL Number: MFCD00147970
CAS Number: 25322-68-3
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty