Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Fluorobenzoyl)propionic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

139900-25G 139900-5G
BT139900-25GEA 79 USD InStock
BT139900-25G BT139900-5G
3-(4-Fluorobenzoyl)propionic acid ≥98%
3-(4-Fluorobenzoyl)propionic acid
CAS: 366-77-8; EC No: 206-679-1; MDL No: MFCD00002793 Solid; Molecular Formula: C10H9FO3; MW: 196.18 Melting Point: 100-102°

  • Solid


Formula: C₁₀H₉FO₃
MW: 196.18 g/mol
Melting Pt: 100…102 °C
MDL Number: MFCD00002793
CAS Number: 366-77-8
EINECS: 206-679-1
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty