Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dinitrosalicylic acid ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

140115-1KG 140115-250G 140115-50G
BT140115-1KGEA 474 USD InStock
BT140115-1KG BT140115-250G BT140115-50G
3,5-Dinitrosalicylic acid ≥97%
3,5-Dinitrosalicylic acid
CAS: 609-99-4; EC No: 210-204-3; MDL No: MFCD00007104; RTECS: VO2965000 Solid; Molecular Formula: C7H4N2O7; MW: 228.12 Melting Point: 168-172°

  • RTECS: VO2965000
  • Solid


Formula: C₇H₄N₂O₇
Melting Pt: 168…172 °C
MDL Number: MFCD00007104
CAS Number: 609-99-4
EINECS: 210-204-3
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty