Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-Methyl-3-hydroxybutyrate ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

140465-1G 140465-25G 140465-5G
BT140465-1GEA 34 USD InStock
BT140465-1G BT140465-25G BT140465-5G
(S)-(+)-Methyl-3-hydroxybutyrate ≥98%
(S)-(+)-Methyl-3-hydroxybutyrate
CAS: 53562-86-0; EC No: 258-628-8; MDL No: MFCD00064461 Liquid; Molecular Formula: C5H10O3; MW: 118.13 Boiling Point: 63°/10 mmHg; Flash point: 73°C (163°F) Density (g/mL): 1.071; Refractive Index: 1.421; Optical Rotation: [α]20/D +19.8°, neat

  • Optical rotation: [α]20/D +19.8°, neat
  • Refractive index: 1.421
  • Liquid


Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 63 °C/10 mmHg
Density: 1.071 g/mL
Flash Pt: 73 °C (163 °F)
MDL Number: MFCD00064461
CAS Number: 53562-86-0
EINECS: 258-628-8
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty