Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl pyrocarbonate ≥97%

Supplier: BEANTOWN CHEMICAL - BTC

140775-100G 140775-25G 140775-5G
BT140775-100GEA 278.69 USD InStock
BT140775-100G BT140775-25G BT140775-5G
Diethyl pyrocarbonate ≥97%
Diethyl pyrocarbonate
CAS: 1609-47-8; EC No: 216-542-8; MDL No: MFCD00009106; RTECS: LQ9350000 Liquid; Molecular Formula: C6H10O5; MW: 162.14 Boiling Point: 93-94°/18 mmHg; Flash point: 69°C (156°F) Density (g/mL): 1.101; Refractive Index: 1.398 Moisture Sensitive

  • RTECS: LQ9350000
  • Refractive index: 1.398
  • Moisture Sensitive
  • Liquid


Formula: O(COOC₂H₅)₂
MW: 162.14 g/mol
Boiling Pt: 93
Density: 1.101 g/mL
Flash Pt: 69 °C (156 °F)
MDL Number: MFCD00009106
CAS Number: 1609-47-8
EINECS: 216-542-8
Merck Index: 13,08088
Order Now
Specification Test Results
Assay 97%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty