Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Titanium ≥99.9% (trace metals basis), foil, hard type, Thickness: 0.025 mm

Supplier: BEANTOWN CHEMICAL - BTC

141050-150X150 141050-25X75MM 141050-75X75MM
BT141050-150X1EA 1041 USD InStock
BT141050-150X1 BT141050-25X75 BT141050-75X75
Titanium ≥99.9% (trace metals basis), foil, hard type, Thickness: 0.025 mm
Titanium
CAS: 7440-32-6; EC No: 231-142-3; MDL No: MFCD00011264; RTECS: XR1700000 Solid; Molecular Formula: Ti; MW: 47.87 Melting Point: 1660°; Boiling Point: 3287° Density (g/mL): 4.5

  • RTECS: XR1700000
  • Solid


Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.5 g/mL
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty