Silver, gauze woven from wire 50 mesh Ø 0.0764 mm

Supplier: BEANTOWN CHEMICAL - BTC

141130-150X150 141130-300X300 141130-75X75MM
BT141130-150X1EA 427 USD
BT141130-150X1 BT141130-300X3 BT141130-75X75
Silver, gauze woven from wire 50 mesh Ø 0.0764 mm
Silver

CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397; RTECS: VW3500000 Wire Cloth ; Molecular Formula: Ag; MW: 107.87 Melting Point: 960°; Boiling Point: 2212° Density (g/mL): 10.49


  • RTECS: VW3500000
  • Wire Cloth

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.49 g/mL
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR