Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(1,3-Dioxoisoindolin-2-yl)propanoic acid ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

141310-1G 141310-5G
BT141310-1GEA 37 USD InStock
BT141310-1G BT141310-5G
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid ≥98%
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid
CAS: 3339-73-9; EC No: 222-083-4; MDL No: MFCD00023096 Solid; Molecular Formula: C11H9NO4; MW: 219.20 Melting Point: 151-153°

  • Solid


Formula: C₁₁H₉NO₄
MW: 219.196 g/mol
Melting Pt: 151…153 °C
MDL Number: MFCD00023096
CAS Number: 3339-73-9
EINECS: 222-083-4
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty