Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tin(II) oxide ≥99.9%

Supplier: BEANTOWN CHEMICAL - BTC

141740-100G 141740-25G 141740-500G
BT141740-100GEA 42.57 USD InStock
BT141740-100G BT141740-25G BT141740-500G
Tin(II) oxide ≥99.9%
Tin(II) oxide
CAS: 21651-19-4; EC No: 244-499-5; MDL No: MFCD00011243; RTECS: XQ3700000 -100 Mesh Powder; Linear Formula: SnO2; MW: 134.69 Melting Point: 1080° (decomposes) Density (g/mL): 6.45 Air Sensitive

  • RTECS: XQ3700000
  • Air Sensitive
  • -100 Mesh Powder


Formula: OSn
MW: 134.71 g/mol
Boiling Pt: 1425
Melting Pt: 1040 (dec.)
Density: 6.45 g/mL
MDL Number: MFCD00011243
CAS Number: 21651-19-4
EINECS: 244-499-5
Merck Index: 12,08944
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty