Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Molybdenum ≥99.95% (trace metals basis), rods Ø 12.7 mm

Supplier: BEANTOWN CHEMICAL - BTC

141795-250G 141795-50G
BT141795-250GEA 228 USD InStock
BT141795-250G BT141795-50G
Molybdenum ≥99.95% (trace metals basis), rods Ø 12.7 mm
Molybdenum
CAS: 7439-98-7; EC No: 231-107-2; MDL No: MFCD00003465 Solid; Molecular Formula: Mo; MW: 95.94 Melting Point: 2617°; Boiling Point: 4612° Density (g/mL): 10.3

  • Solid


Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
Density: 10.3 g/mL
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317
Order Now
Specification Test Results
Assay 99.95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty