Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Platinum ≥99.95% (trace metals basis), wire Ø 0.5 mm, annealed

Supplier: BEANTOWN CHEMICAL - BTC

141850-100CM 141850-25CM
BT141850-100CMEA 969.28 USD InStock
BT141850-100CM BT141850-25CM
Platinum ≥99.95% (trace metals basis), wire Ø 0.5 mm, annealed
Platinum
CAS: 7440-06-4; EC No: 231-116-1; MDL No: MFCD00011179; RTECS: TP2160000 Solid; Molecular Formula: Pt; MW: 195.08 Melting Point: 1772°; Boiling Point: 3827° Density (g/mL): 21.45

  • RTECS: TP2160000
  • Solid


Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.45 g/mL
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689
Order Now
Specification Test Results
Assay 99.95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty