Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Titanium nitride ≥99.5%

Supplier: BEANTOWN CHEMICAL - BTC

142420-1KG 142420-250G 142420-50G
BT142420-1KGEA 650 USD InStock
BT142420-1KG BT142420-250G BT142420-50G
Titanium nitride ≥99.5%
Titanium nitride
CAS: 25583-20-4; EC No: 247-117-5; MDL No: MFCD00049596; RTECS: XR2230000 -325 Mesh Powder; Linear Formula: TiN; MW: 61.89 Melting Point: 2930° Density (g/mL): 5.24

  • RTECS: XR2230000
  • -325 Mesh Powder


Formula: NTi
MW: 61.89 g/mol
Melting Pt: 2930
Density: 5.24 g/mL
MDL Number: MFCD00049596
CAS Number: 25583-20-4
EINECS: 247-117-5
Order Now
Specification Test Results
Assay 99.5%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty