Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-(+)-Galactosamine hydrochloride ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

142730-1G 142730-250MG
BT142730-1GEA 92.32 USD InStock
BT142730-1G BT142730-250MG
D-(+)-Galactosamine hydrochloride ≥98%
D(+)-Galactosamine hydrochloride
CAS: 1772-03-8; EC No: 217-198-1; MDL No: MFCD00135830; RTECS: LW5500000 Powder; Molecular Formula: C6H13NO5·HCl; MW: 215.64 Hygroscopic

  • RTECS: LW5500000
  • Hygroscopic
  • Powder


Formula: C₆H₁₄ClNO₅
MW: 215.64 g/mol
MDL Number: MFCD00135830
CAS Number: 1772-03-8
EINECS: 217-198-1
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty