142900-100G
142900-25G
142900-5G
BT142900-100GEA
1035
USD
InStock
BT142900-100G
BT142900-25G
BT142900-5G
1-Bromo-3-chloro-5-fluorobenzene ≥98%
1-Bromo-3-chloro-5-fluorobenzene
CAS: 33863-76-2; MDL No: MFCD00070747
Liquid; Molecular Formula: C6H3BrClF; MW: 209.45
Density (g/mL): 1.72