Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nickel chromite

Supplier: BEANTOWN CHEMICAL - BTC

143125-100G 143125-25G
BT143125-100GEA 693 USD InStock
BT143125-100G BT143125-25G
Nickel chromite
Nickel chromite
CAS: 12018-18-7; EC No: 234-636-7; MDL No: MFCD00064806; RTECS: GB9000000 -100 Mesh Granular; Molecular Formula: Cr2NiO4; MW: 226.70

  • RTECS: GB9000000
  • Granular


Formula: Cr₂NiO₄
MDL Number: MFCD00064806
CAS Number: 12018-18-7
EINECS: 234-636-7
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty