Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Cyclohexene-1-acetic acid

Supplier: BEANTOWN CHEMICAL - BTC

143330-1G 143330-25G 143330-5G
BT143330-1GEA 30 USD InStock
BT143330-1G BT143330-25G BT143330-5G
1-Cyclohexene-1-acetic acid
1-Cyclohexene-1-acetic acid
CAS: 18294-87-6; EC No: 242-172-1; MDL No: MFCD00015482 Liquid/Low Melting Solid; Molecular Formula: C8H12O2; MW: 140.18 Melting Point: 32-35° Density (g/mL): 1.04

  • Liquid/Low Melting Solid


Formula: C₈H₁₂O₂
MW: 140.182 g/mol
Melting Pt: 32…35 °C
Density: 1.04 g/mL
MDL Number: MFCD00015482
CAS Number: 18294-87-6
EINECS: 242-172-1
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty