143370-100G
143370-500G
BT143370-100GEA
65
USD
InStock
BT143370-100G
BT143370-500G
Butyramide ≥98%
Butyramide
CAS: 541-35-5; EC No: 208-776-4; MDL No: MFCD00041894
Powder/Flakes; Linear Formula: CH3CH2CH2CONH2; Molecular Formula: C4H9NO; MW: 87.12
Melting Point: 114-116°