143730-100G
143730-25G
143730-500G
BT143730-100GEA
474
USD
InStock
BT143730-100G
BT143730-25G
BT143730-500G
Iminostilbene ≥97%
Iminostilbene
CAS: 256-96-2; EC No: 205-970-0; MDL No: MFCD00005071
Powder; Molecular Formula: C14H11N; MW: 193.25
Melting Point: 196-199°