Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Thiopheneethanol ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

143850-25G 143850-5G
BT143850-25GEA 167 USD InStock
BT143850-25G BT143850-5G
2-Thiopheneethanol ≥98%
2-Thiopheneethanol
CAS: 5402-55-1; EC No: 226-452-0; MDL No: MFCD00005462 Liquid; Molecular Formula: C6H8OS; MW: 128.19 Boiling Point: 107-108°/12 mmHg; Flash point: 101°C (213°F) Density (g/mL): 1.150; Refractive Index: 1.551

  • Refractive index: 1.551
  • Liquid


Formula: C₆H₈OS
MW: 128.19 g/mol
Boiling Pt: 107…108 °C/12 mmHg
Density: 1.150 g/mL
Flash Pt: 101 °C (213 °F)
MDL Number: MFCD00005462
CAS Number: 5402-55-1
EINECS: 226-452-0
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty