Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chlorophenol red sodium salt 0,04% (w/v) in water

Supplier: BEANTOWN CHEMICAL - BTC

143950-100ML 143950-500ML
BT143950-100MLEA 27 USD InStock
BT143950-100ML BT143950-500ML
Chlorophenol red sodium salt 0,04% (w/v) in water
Chlorophenol red sodium salt
CAS: 123333-64-2; MDL No: MFCD00151199 Liquid; Molecular Formula: C19H11Cl2NaO5S; MW: 445.26 Density (g/mL): 0.99

  • Liquid


Formula: C₁₉H₁₂Cl₂O₅S
MW: 445.24 g/mol
Density: 0.99 g/mL
MDL Number: MFCD00151199
CAS Number: 123333-64-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty