144070-10G
144070-50G
BT144070-10GEA
81
USD
InStock
BT144070-10G
BT144070-50G
5-Chloro-1-pentanol 95
5-Chloro-1-pentanol
CAS: 5259-98-3; EC No: 226-067-8; MDL No: MFCD00039556; RTECS: KO1100000
Liquid; Linear Formula: Cl(CH2)5OH; MW: 122.60
Boiling Point: 102-103°/8 mmHg
Density (g/mL): 1.06