Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lutetium(III) oxide ≥99.9%

Supplier: BEANTOWN CHEMICAL - BTC

144385-1G 144385-25G 144385-5G
BT144385-1GEA 46 USD InStock
BT144385-1G BT144385-25G BT144385-5G
Lutetium(III) oxide ≥99.9%
Lutetium(III) oxide
CAS: 12032-20-1; EC No: 234-764-3; MDL No: MFCD00011100 Powder; Molecular Formula: Lu2O3; MW: 397.94 Density (g/mL): 9.42 Hygroscopic

  • Hygroscopic
  • Powder


Formula: Lu₂O₃
MW: 397.93 g/mol
Melting Pt: 2510
Density: 9.42 g/mL
MDL Number: MFCD00011100
CAS Number: 12032-20-1
EINECS: 234-764-3
Order Now
Specification Test Results
Assay 99.9%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty