Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium molybdate(VI) ≥95%

Supplier: BEANTOWN CHEMICAL - BTC

144915-100G 144915-500G
BT144915-100GEA 68 USD InStock
BT144915-100G BT144915-500G
Potassium molybdate(VI) ≥95%
Potassium molybdate(VI)
CAS: 13446-49-6; EC No: 236-599-2; MDL No: MFCD00011370 Powder; Molecular Formula: K2MoO4; MW: 238.14 Melting Point: 919° Density (g/mL): 2.3 Hygroscopic

  • Hygroscopic
  • Powder


Formula: K₂MoO₄
Melting Pt: 919 °C
Density: 2.3 g/mL
MDL Number: MFCD00011370
CAS Number: 13446-49-6
EINECS: 236-599-2
Order Now
Specification Test Results
Assay 95%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty