Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethyl-3-methylimidazolium bromide ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

144970-10G 144970-50G
BT144970-10GEA 53 USD InStock
BT144970-10G BT144970-50G
1-Ethyl-3-methylimidazolium bromide ≥98%
1-Ethyl-3-methylimidazolium bromide
CAS: 65039-08-9; MDL No: MFCD03427610 Crystalline; Molecular Formula: C6H11BrN2; MW: 191.07

  • Crystalline


Formula: C₆H₁₁BrN₂
MDL Number: MFCD03427610
CAS Number: 65039-08-9
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty