Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Riboflavine (vitamin B2) ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

144995-100G 144995-25G
BT144995-100GEA 71.09 USD InStock
BT144995-100G BT144995-25G
Riboflavine (vitamin B2) ≥98%
Riboflavine (vitamin B2)
CAS: 83-88-5; EC No: 201-507-1; MDL No: MFCD00005022 Powder; Molecular Formula: C17H20N4O6; MW: 376.37 Melting Point: 290° (decomposes)

  • Powder


Formula: C₁₇H₂₀N₄O₆
MW: 376.37 g/mol
Melting Pt: 280 (dec.)
MDL Number: MFCD00005022
CAS Number: 83-88-5
EINECS: 201-507-1
Merck Index: 12,08367
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty