Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bismuth(III) 2-ethylhexanoate

Supplier: BEANTOWN CHEMICAL - BTC

145090-25G 145090-5G
BT145090-100GEA 264.55 USD InStock
BT145090-100G BT145090-25G BT145090-5G
Bismuth(III) 2-ethylhexanoate
Bismuth (III) 2-ethylhexanoate
CAS: 67874-71-9; EC No: 267-499-7; MDL No: MFCD00043803 Liquid; Molecular Formula: C24H45BiO6; MW: 638.61

  • Liquid


Formula: C₂₄H₄₅BiO₆
MW: 638.598 g/mol
MDL Number: MFCD00043803
CAS Number: 67874-71-9
EINECS: 267-499-7
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty