Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione ≥98%

Supplier: BEANTOWN CHEMICAL - BTC

145240-25G 145240-5G
BT145240-25GEA 89.5 USD InStock
BT145240-25G BT145240-5G
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione ≥98%
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione
CAS: 4511-42-6; EC No: 224-832-0; MDL No: MFCD00070594 Solid; Molecular Formula: C6H8O4; MW: 144.13 Melting Point: 92-94° Optical Rotation: [α]20/D -285°, c = 1 in toluene Moisture Sensitive

  • Optical rotation: [α]20/D -285°, c = 1 in toluene
  • Moisture Sensitive
  • Solid


Formula: C₆H₈O₄
MW: 144.13 g/mol
Melting Pt: 92…94 °C
MDL Number: MFCD00070594
CAS Number: 4511-42-6
EINECS: 224-832-0
Order Now
Specification Test Results
Assay 98%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty