Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methoxy(cyclooctadiene)iridium(I) dimer

Supplier: BEANTOWN CHEMICAL - BTC

145605-1G 145605-5G
BT145605-1GEA 361 USD InStock
BT145605-1G BT145605-5G
Methoxy(cyclooctadiene)iridium(I) dimer
Methoxy(cyclooctadiene)iridium(I) dimer
CAS: 12148-71-9; MDL No: MFCD08459360 Crystalline/Powder; Linear Formula: [Ir(OCH3)(C8H12)]2; Molecular Formula: C18H30Ir2O2; MW: 662.88 Melting Point: 154-179° (decomposes) Air Sensitive, Light Sensitive, Moisture Sensitive

  • Air Sensitive, Light Sensitive, Moisture Sensitive
  • Crystalline/Powder


Formula: C₁₈H₃₀Ir₂O₂
MW: 662.87 g/mol
Melting Pt: 154…179 °C (dec.)
MDL Number: MFCD08459360
CAS Number: 12148-71-9
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty