Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium thiophosphate hydrate

Supplier: BEANTOWN CHEMICAL - BTC

145665-25G 145665-5G
BT145665-25GEA 292 USD InStock
BT145665-25G BT145665-5G
Sodium thiophosphate hydrate
Sodium thiophosphate hydrate
CAS: 10489-48-2; MDL No: MFCD00041513 Solid; Linear Formula: Na3PO3S·xH2O; MW: 180.00 (anhydrous) Melting Point: 53-58°

  • Solid


Formula: H₂Na₃O₄PS
MDL Number: MFCD00041513
CAS Number: 10489-48-2
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty