Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrite (FeS₂) naturally occurring mineral 7.6x7.6 cm

Supplier: BEANTOWN CHEMICAL - BTC

145705-1PC 145705-5PC
BT145705-1PCEA 179 USD InStock
BT145705-1PC BT145705-5PC
Pyrite (FeS₂) naturally occurring mineral 7.6x7.6 cm
Pyrite (FeS₂)
CAS: 1309-36-0; EC No: 215-167-7; MDL No: MFCD00064690 Solid; Molecular Formula: FeS2; MW: 119.98 Density (g/mL): 5.02

  • Solid


Formula: FeS₂
MW: 119.965 g/mol
Density: 5.02 g/mL
MDL Number: MFCD00064690
CAS Number: 1309-36-0
EINECS: 215-167-7
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty