Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper ≥99.8% (trace metals basis), foil, annealed, coated, Thickness: 0.025 mm

Supplier: BEANTOWN CHEMICAL - BTC

145780-30X1000 145780-30X150 145780-30X30CM
BT145780-30X10EA 134 USD InStock
BT145780-30X10 BT145780-30X15 BT145780-30X30
Copper ≥99.8% (trace metals basis), foil, annealed, coated, Thickness: 0.025 mm
Copper
CAS: 7440-50-8; EC No: 231-159-6; MDL No: MFCD00010965; RTECS: GL5325000 Solid; Molecular Formula: Cu; MW: 63.55 Melting Point: 1083.4°; Boiling Point: 2567° Density (g/mL): 8.94

  • RTECS: GL5325000
  • Solid


Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/mL
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545
Order Now
Specification Test Results
Assay 99.8%

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty